Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Mouse, iFluor 647, Clone: 6G2A9, THE™, GenScript™

Catalog No. 502120594
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0594 Supplier Genscript Corporation Supplier No. A01802100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. The His tag is a short amino acid sequence consists of Histidine (His) residues appended to recombinant proteins. Ni-charged resins are used for the purification of His-tagged proteins from eukaryotic and prokaryotic expression systems. An anti-His tag antibody is a useful tool for the analysis of His-tagged proteins.THE™ His Tag Antibody [iFluor 647], mAb, Mouse is THE™ His Tag Antibody (A00186) conjugated with iFluor 647 under optimal conditions with an F/P ratio of 3-6. It is suitable for detecting the expression level of His-tagged proteins. iFluor 647 is a bright and photostable fluorescent dye. It is an excellent alternative to Alexa Fluor 647.

His Tag Monoclonal antibody specifically detects His Tag in Tag samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen His Tag
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 6G2A9
Conjugate iFluor 647
Formulation PBS with 10mg/mL BSA and 0.02% sodium azide; pH 7.4
Gene Alias 6His tag; 6X His; His Tag; Histidine Tag
Host Species Mouse
Immunogen A synthetic peptide HHHHHH coupled to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.