Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Rabbit anti-All, Clone: 6HIS/6402R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen His Tag
Clone 6HIS/6402R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunoprecipitation
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122016
View Documents
Novus Biologicals
NBP30727120UG
20 μg
Each for $159.82
Add to cart
 
NB122015
View Documents
Novus Biologicals
NBP30727110UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

His Tag Monoclonal antibody specifically detects His Tag in All samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

His Tag
0.2 mg/ml
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Purified
RUO
All
6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag
IgG Kappa
Primary
Store at 4°C.
6HIS/6402R
Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunoprecipitation
Unconjugated
Rabbit
Cellular Markers, Epitope Tags
10 mM PBS with 0.05% BSA
Hexa-histidine tagged human CD70 recombinant protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit