Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His-tag Rabbit anti-All Species, Biotin, Clone: 146, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262177B

Catalog No. NBP262177B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

His-tag
146
Biotin
50% Glycerol/PBS with 1% BSA
Rabbit
IgG
Store at -20°C.
Monoclonal
All
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
1mg/mL
Western Blot 0.1μg/mL-0.5μg/mL, Flow Cytometry 0.5μg/mL-2μg/mL, ELISA 0.01μg/mL-0.5μg/mL, Immunohistochemistry 0.1μg/mL-1μg/mL, Immunocytochemistry 0.5μg/mL-2μg/mL
Purified
Mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis-Tag at the C-terminus.
50μg
Primary
This antibody reacts to recombinant proteins containing the 6xHis-Tag or 10xHis-Tag fused to either the amino or carboxy terminus. No cross reactivity with other endogenous protein in mammalian or bacteria cells.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit