Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag Rabbit anti-Human, Non-species specific, Alexa Fluor™ 350, Clone: 3D5, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252637AF350

Catalog No. NP22637A30


Add to cart

Description

Description

His Tag Monoclonal antibody specifically detects His Tag in Human,Non-species specific samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

His Tag
3D5
Western Blot, ELISA
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Purified his tagged Fab fragment.
0.1 mL
Primary
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags.
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Human, Nonspecies specific
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit