Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His Tag Antibody (3D5), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252637AF750
Description
His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.Specifications
His Tag | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
Rabbit | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
Epitope Tag |
Western Blot, ELISA | |
3D5 | |
Western Blot, ELISA | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
Purified his tagged Fab fragment. | |
Cellular Markers, Epitope Tags | |
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags. | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction