Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
His tag Rabbit anti-Tag, liquid, TrueRAB™

Rabbit Recombinant Monoclonal Antibody
Supplier: OriGene TA591008S

Description
Synthetic peptide sequence is HHHHHH
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombit proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombit protein using genetic engineering. By choosing a particular epitope and recombit protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombit proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent.Specifications
His tag | |
Recombinant Monoclonal | |
Unconjugated | |
ANTI HIS, ANTI HISTIDINE, ANTIHIS, anti-His, his tag antibody, Anti-His Antibody, his antibody, anti his tag, anti-his tag | |
Synthetic peptide | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
OR-5B12 | |
Tissue Culture Supernatant with 1% BSA and 0.02% sodium azide | |
Rabbit | |
30 μL | |
Primary | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction