Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

His Tag, Rat anti-Non-species specific, Clone: 3D5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP2810230.2MG

 View more versions of this product

Catalog No. NB014202


Add to cart

Description

Description

His Tag Monoclonal antibody specifically detects His Tag in Non-species specific Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

His Tag
3D5
Unconjugated
PBS
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Purified his tagged Fab fragment.
Protein A purified
RUO
Primary
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags.
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA
Purified
Rat
IgG2a Kappa
0.2 mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Nonspecies specific
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit