Learn More
GE Healthcare HiScale™ Chromatography Columns: PROMO
Product
Manufacturer:  GE Healthcare 28964514PM
 View more versions of this product
Catalog No. 45-200-32LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Empty columns with high pressure stability for packing with modern BioProcess media for process development and preparative chromatography.
  • Provides axial compression of the gel bed and a nonrotating plunger mechanism that expands packing capabilities.
  • Adapter QuickLock mechanism, column measurement scale, and ergonomic design, facilitate column handling and increase process control.
  • Dual adapters, offering high flexibility and dynamic packing heights.
  • Capability to rinse entire adapter shaft after packing, thereby minimizing risk of bacterial growth after prolonged use.
  • Based on the well-established line of XK columns and compatible with all methods currently applicable to XK columns.
Specifications
Specifications
106mL
HiScale Chromatography Column
200mm
Borosilicate glass
Polypropylene (reinforced glassfibre)/Polyamide Net
26mm
Process development and preparative chromatography
Column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors
20 bar