Learn More
GE Healthcare HiScale™ Chromatography Columns: PROMO
Product
Specifications
Support Type Polypropylene (reinforced glassfibre)/Polyamide Net
Max. Pressure 20 bar
Includes Column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors
Material Borosilicate glass
View More Specs
Products 3
Catalog Number Mfr. No. Bed Volume Height (Metric) Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Bed Volume Height (Metric) Diameter (Metric) Inner Price Quantity & Availability  
45-200-29LR View Documents GE Healthcare
28964424PM
80mL 400mm 16mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-200-32LR View Documents GE Healthcare
28964514PM
106mL 200mm 26mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
45-200-30LR View Documents GE Healthcare
28964445PM
393mL 200mm 50mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Empty columns with high pressure stability for packing with modern BioProcess media for process development and preparative chromatography.
  • Provides axial compression of the gel bed and a nonrotating plunger mechanism that expands packing capabilities.
  • Adapter QuickLock mechanism, column measurement scale, and ergonomic design, facilitate column handling and increase process control.
  • Dual adapters, offering high flexibility and dynamic packing heights.
  • Capability to rinse entire adapter shaft after packing, thereby minimizing risk of bacterial growth after prolonged use.
  • Based on the well-established line of XK columns and compatible with all methods currently applicable to XK columns.
Specifications
Specifications
Polypropylene (reinforced glassfibre)/Polyamide Net
20 bar
Column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors
Borosilicate glass