Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ Q ImpRes and Capto™ SP ImpRes

High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands

$512.50

Specifications

Buffer 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Diameter (English) Inner 39.3 in.
Diameter (Metric) Inner 1000mm
Quantity 100mL
Flow Rate 220cm/hr.
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Ligand Type pH Range Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Ligand Type pH Range Price Quantity & Availability  
45-002-562
View Documents Promotion Details
Cytiva
17546802
Capto SP ImpRes medium Sulphonate group 4 to 12
Each for $512.50
Add to cart
 
45-002-564
View Documents Promotion Details
Cytiva
17547002
Capto Q ImpRes medium Quaternary Amine 2 to 12
Each for $512.50
Add to cart
 
Description

Description

  • Delivers pressure-flow properties with wide resolution
  • High-throughput purifications easy to optimize and scale up
  • High flow agarose based matrix
  • Runs at high flow rates as well as high bed heights thus increasing flexibility in process design
  • Developed for large-scale purification of biopharmaceutical proteins
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Available in a wide range of different bulk pack sizes and pre-packed formats for process development

Capto Q ImpRes: Strong anion (quaternary amine group) exchange BioProcess™ medium
Capto SP ImpRes: Strong cation (sulfonate group) exchange BioProcess medium

Specifications

Specifications

1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
1000mm
220cm/hr.
7.87 in.
<3 bar
4° to 30°C
39.3 in.
100mL
High-throughput intermediate purification and polishing steps of a wide range of biomolecules
200mm
36 to 44μm
High-flow Agarose
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit