Learn More
Seracare Life Sciences IncSupplier Diversity Partner HISDETECTOR NICKEL-AP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 58200003
HisDetector Nickel-AP offers a fast, sensitive, single-step method for detecting recombinant His-tagged proteins. His-tagged proteins have a high affinity to nickel (Ni2+) ions. Nickel-NTA (nitrilotriacetic acid) conjugated to alkaline phosphatase(AP) will therefore bind directly to the His-tag, eliminating the need for primary and secondary antibodies. This results in assay simplification and reduced detection times. Nickel-AP offers excellent sensitivity and low background in applications such as Western Blotting (see KPL Application Note). Product may also be used in other applications (ELISA, immunohistochemistry, etc.) where direct detection of His-tagged proteins is required. Nickel-AP is optimized for use with both chemiluminescent and colorimetric substrates.
Catalog No. 50-673-82