Learn More
Seracare Life Sciences IncSupplier Diversity Partner HISDETECTOR WB KIT HRP COLOR  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 58100023
HisDetector Western Blot Kit, HRP Colorimetric uses Nickel-HRP and TMB colorimetric substrate to produce a deep purple band at the site of reaction. Kit utilizes the specificity and high affinity of Nickel Conjugates (read more about Nickel conjugate technology) and optimized reagents to provide picogram level sensitivity with low background. See our Application Note for more information on the performance of HisDetector Western Blot Kits. Sufficient reagents provided for running 40 blots (10 x 10 cm2 blots)
Catalog No. 50-673-78