Learn More
Thermo Scientific™ HisPur™ Ni-NTA Chromatography Cartridges, 1 mL
Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.
$155.65 - $340.50
Specifications
Content And Storage | Store between 4°C and 8°C. Do not freeze. |
---|---|
Product Line | HisPur™ |
Target | His-Tagged Protein |
Type | Chromatography Cartridge |
Stationary Phase | Divalent Nickel, Metal Chelate |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
PI90098
|
Thermo Scientific™
90098 |
HisPur Ni-NTA Chromatography Cartridges, 1mL | 5 Cartridges |
Pack of 5
for $155.65
Save 21% Reg : |
|
||||
PI90099
|
Thermo Scientific™
90099 |
HisPur Ni-NTA Chromatography Cartridges, 5mL | 2 x 5 mL |
Pack of 2 for $340.50
|
|
||||
Description
Perform routine affinity purification of His-tagged fusion proteins with Thermo Scientific™ HisPur Ni-NTA Resin, a high-capacity, high-performance nickel-IMAC resin.
The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.
Highlights:
High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
Versatile – purify proteins using native or denaturing conditions
Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
Cost-effective – reuse the same batch of resin at least five times
Easy to use – pre-formulated buffers available for kit formats
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
- Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin
Includes:
Kits contain HisPur Ni-NTA Columns
Specifications
Store between 4°C and 8°C. Do not freeze. | |
His-Tagged Protein | |
Divalent Nickel, Metal Chelate |
HisPur™ | |
Chromatography Cartridge | |
Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.