Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H2BN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548515

Catalog No. PA548515


Add to cart

Description

Description

Predicted to react with mouse and bovine based on sequence homology.

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications

Specifications

HIST1H2BN
Polyclonal
PBS with 0.09% sodium azide
Q99877
Histone H2B type 1-N, Histone H2Bd, H2B/d, HIST1H2BN, H2BFD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8341
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HIST1H2BN
Liquid
HIST1H2BN
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HIST1H2BN.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit