Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A (p Ser10) Mouse anti-Human, Rat, DyLight 550, Clone: 921, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243620R

Catalog No. NBP243620R


Add to cart

Description

Description

HIST1H3A p Ser10 Monoclonal antibody specifically detects HIST1H3A p Ser10 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

HIST1H3A (p Ser10)
921
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
HIST1H3A
The exact sequence is proprietary.
Protein G purified
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors
Primary
8350
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
H3 histone family, member A, H3/A, H3FAHIST1H3E, H3FB, H3FC, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3a, histone cluster 1, H3a, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit