Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3D (dimethyl-K27) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against synthetic Di-methyl-peptide of human HIST1H3D.

Manufacturer:  Abnova Corporation PAB29623

Catalog No. 89-947-455


Add to cart

Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq]

Specifications

Specifications

HIST1H3D (dimethyl-K27)
Unconjugated
Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Liquid
HIST1H3D
A synthetic Di-methyl-peptide (conjugated with KLH) corresponding to residues surrounding K27 of human HIST1H3D.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rabbit
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic Di-methyl-peptide of human HIST1H3D.
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
H3/b|H3FB
Rabbit
Affinity Chromatography
RUO
Primary
8351
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit