Learn More
HIST1H3D (trimethyl-K27) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic Tri-methyl-peptide of human HIST1H3D.
Manufacturer:  Abnova Corporation PAB29622
Catalog No. 89-947-457
Add to cart
Description
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq]
Specifications
Specifications
HIST1H3D (trimethyl-K27)
Unconjugated
Immunofluorescence (1:100-1:200) Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Liquid
HIST1H3D
A synthetic Tri-methyl-peptide (conjugated with KLH) corresponding to residues surrounding K27 of human HIST1H3D.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rabbit
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic Tri-methyl-peptide of human HIST1H3D.
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
H3/b|H3FB
Rabbit
Affinity Chromatography
RUO
Primary
8351