Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H4A Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: HDC-8, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20165
Description
HIST1H4A Monoclonal antibody specifically detects HIST1H4A in Human, Mouse, Rat samples. It is validated for WB, IP, ChIP, ICC, IHC-P, IF, Flow Cyt.Specifications
HIST1H4A | |
Monoclonal | |
Unconjugated | |
HIST1H4A | |
Rabbit | |
100 μg | |
8359 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
HDC-8 | |
histone cluster 1, H4a | |
H4/a, H4FA | |
A synthetic peptide corresponding to human HIST1H4A | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction