Learn More
Aviva Systems Biology Corp HIST2H2AC Antibody - middle region (ARP51480_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51480_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HIST2H2AC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIST2H2AC. Alternate names: H2A, H2A-GL101, H2A/q, H2AFQ, MGC74460ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60567