Learn More
Histamine H1R Mouse anti-Human, Alexa Fluor 700, Clone: 480054, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4726N100UG
Catalog No. FAB4726N100
Add to cart
This item is not returnable. View return policy
Description
Description
Histamine H1R Monoclonal specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Histamine H1R
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
AAH60802
HRH1
NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Histamine H1 R. Stains human Histamine H1 R transfectants but not irrelevant transfectants.
Flow Cytometry
480054
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
H1R, H1-R, hisH1, Histamine H1R, histamine receptor H1, HRH1, subclass H1
Mouse
IgG1
RUO
Primary
3269
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)