Learn More
Histamine N-Methyltransferase/HNMT Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300070UV
Catalog No. NB064240
Add to cart
Description
Description
Histamine N-Methyltransferase/HNMT Polyclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Histamine N-Methyltransferase/HNMT
DyLight 350
50mM Sodium Borate
EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2
Produced in rabbits immunized with purified, recombinant Histamine N-Methyltransferase/HNMT (AAH20677.1; Met 1-Ala 292).
Affinity purified
RUO
Primary
3176
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human