Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine N-Methyltransferase/HNMT Rabbit anti-Human, DyLight 488, Clone: 34, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290171G

Catalog No. NB037864


Add to cart

Description

Description

Histamine N-Methyltransferase/HNMT Monoclonal antibody specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Histamine N-Methyltransferase/HNMT
34
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 488
50mM Sodium Borate
EC 2.1.1.8, histamine N-methyltransferase, HMT, HNMT-S1, HNMT-S2
Recombinant Human Histamine N-Methyltransferase/HNMT protein
Protein A purified
RUO
Primary
3176
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit