Learn More
3B Scientific™ Histology of Mammalia Elementary Slide Set
25 microscope slides
Supplier: 3B Scientific™ W13406
Description
- 1(c). Squamous epithelium, isolated cells
- 2(e). Fibrous connective tissue, w.m. from pig mesentery
- 3(e). Adipose tissue of mammal, fat stained
- 4(c). Hyaline cartilage of calf, t.s.
- 5(e). Compact bone of cow, t.s.
- 6(d). Striated muscles of cat, l.s.
- 7(d). Smooth muscles of cat, t.s. and l.s.
- 8(c). Blood smear, human
- 9(d). Artery of cat or rabbit, t.s.
- 10(d). Vein of cat or rabbit, t.s.
- 11(c). Lung of cat, t.s.
- 12(c). Pancreas of pig with islets of Langerhans t.s.
- 13(c). Tongue of cat, t.s. with cornified papillae
- 14(d). Stomach of cat, fundic region t.s.
- 15(c). Small intestine of cat or rabbit, t.s.
- 16(d). Liver of pig, t.s.
- 17(d). Kidney of cat, t.s.
- 18(d). Ovary of rabbit, t.s., developing follicles
- 19(d). Testis of mouse, t.s., spermatogenesis
- 20(d). Cerebrum of cat, t.s.
- 21(d). Cerebellum of cat, t.s.
- 22(c). Spinal cord of cat, t.s.
- 23(e). Nerve fibres isolated, Ranvier's nodes
- 24(e). Motor nerve cells, smear from spinal cord
- 25(d). Scalp, human, l.s. of hair follicles
Specifications
Histology of Mammalia | |
25 |