Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Histology Paraffin Chill Tray Available on GSA/VA Contract for Federal Government customers only.

High performance, well insulated cabinet unit that saves time in the histology laboratory. Mopec™ Histology Paraffin Chill Tray is a mobile, self contained refrigeration unit designed to rapidly cool paraffin embedded tissue samples during histologic processing.

Manufacturer:  Mopec BK105

Catalog No. 22-444-009


Add to cart

Description

Description

  • Constructed of stainless steel
  • Includes 3 inch diameter rubber casters that allow for easy storage and mobility throughout the laboratory

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

1 Year Warranty

Specifications

Specifications

Ambient to -25°C
81.28 cm
55.88 cm
16 in.
32 in.
22 in.
Chill Tray
40.46 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit