Learn More
Seracare Life Sciences IncSupplier Diversity Partner HISTOMARK RED AP SUBSTRATE KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 55100036
The HistoMark RED system contains reagents for the detection of alkaline phosphatase-labeled conjugates. HistoMark RED reagents form a brilliant scarlet reaction product that is stable in organic solvents. Sections reacted with HistoMark RED can be dehydrated through graded alcohols and xylene, then mounted in xylene-based mounting media for permanent documentation. The hematoxylin counterstain, Contrast BLUE, provides sharp contrast and excelllent nuclear detail.
Catalog No. 50-675-29