Learn More
Seracare Life Sciences IncSupplier Diversity Partner HISTOMARKBLUE AP SUBSTRATE KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 55100037
The HistoMark BLUE system contains reagents for the detection of alkaline phosphatase-labeled conjugates. HistoMark BLUE reagents produces a royal blue reaction product that is alcohol soluble, and aqueous mounting media should be used. Contrast RED, a nuclear counterstain formulated from nuclear Fast Red, provides excellent nuclear detail.
Catalog No. 50-675-30