Learn More
Alfa Aesar™ Histone Acetyltransferase Inhibitor IV, CPTH2
Manufacturer:  Alfa Aesar™ J65939MA
 View more versions of this product
Catalog No. AAJ65939MA
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
Specifications
Specifications
10mg
2° to 8°C
A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor
Soluble in DMSO
Brown
Solid
357649-93-5
291.80g/mol
Light Sensitive
Signal Transduction Reagent