Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase Inhibitor III

$118.30 - $227.50

Specifications

CAS 251456-60-7
Formula Weight 307.39g/mol
Synonym M 344; 4-Dimethylamino-N-(6-hydroxycarbamoylhexyl) benzamide
Storage Requirements −30° to −10°C
Format Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64940MA View Documents Alfa Aesar™
J64940MA
10mg Each for $227.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ64940LB0 View Documents Alfa Aesar™
J64940LB0
5mg Each for $118.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

251456-60-7
M 344; 4-Dimethylamino-N-(6-hydroxycarbamoylhexyl) benzamide
Powder
White
307.39g/mol
−30° to −10°C
Light sensitive
Signal Transduction Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit