Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone 1F0 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550984

Catalog No. PA550984


Add to cart

Description

Description

Histone H1.0 / H1F0 Polyclonal Antibody detects endogenous levels of Histone H1.0 / H1F0 protein.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H1 family. [provided by RefSeq, Jul 2008].
Specifications

Specifications

Histone 1F0
Polyclonal
Unconjugated
H1F0
Liquid
H1F0
Synthesized peptide derived from the Internal region of human Histone 1.0.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P07305
H1F0, H1FV, Histone H1.0, Histone H1′, Histone H1(0)
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3005
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit