Learn More
Histone H1.5S17ph Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 61107
Encompass
 View more versions of this product
Catalog No. 50-199-3147
Add to cart
Description
Description
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family.
Specifications
Specifications
Histone H1.5S17ph
Dot Blot, Western Blot
Unconjugated
H1-5
Liquid
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
3009
H1-5
Lot-Specific
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
P16401
H1; H1 histone family; H1 histone family, member 5; H1.5; H1B; H1F5; H1s-3; HIST1H1B; histone 1, H1b; histone cluster 1, H1b; Histone H1.5; Histone H1a; Histone H1b; Histone H1s-3; MGC126630; MGC126632
This Histone H1.5 phospho Ser17 antibody was raised against a peptide containing phospho Ser17 of human histone H1.5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human