Learn More
Histone H1 Mouse anti-Human, Clone: AE-4, Abnova™
Mouse monoclonal antibody raised against native human Histone H1.
Manufacturer:  Abnova Corporation MAB13301
Catalog No. 89-934-244
Add to cart
Description
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H1 family. [provided by RefSeq]
Specifications
Specifications
Histone H1
AE-4
Mouse monoclonal antibody raised against native human Histone H1.
In 1mM PBS (0.05% BSA, 0.05% sodium azide).
H10/H1FV/MGC5241
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
H1F0
Nuclei of human leukemia biopsy cells.
Protein A/G purification
RUO
Primary
3005