Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H1T Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Histone H1T |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1530020
|
Novus Biologicals
NBP15300120UL |
20 μL |
Each for $152.22
|
|
NBP153001
|
Novus Biologicals
NBP153001 |
100 μL |
Each for $436.00
|
|
Description
Histone H1T Polyclonal specifically detects Histone H1T in Human samples. It is validated for Western Blot.Specifications
Histone H1T | |
Polyclonal | |
Rabbit | |
Epigenetics, Histones and Modified Histones | |
P22492 | |
3010 | |
Synthetic peptides corresponding to HIST1H1T(histone cluster 1, H1t) The peptide sequence was selected from the middle region of HIST1H1T. Peptide sequence KLSKKVIPKSTRSKAKKSVSAKTKKLVLSRDSKSPKTAKTNKRAKKPRAT. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
dJ221C16.2, H1 histone family, member T (testis-specific), H1FTMGC163222, H1t, histone 1, H1t, histone cluster 1, H1t, histone H1t, Testicular H1 histone | |
HIST1H1T | |
IgG | |
Affinity Purified | |
22 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title