Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2A/H4S1ph Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
Supplier: Active Motif 39115
Specifications
Histone H2A/H4S1ph | |
Polyclonal | |
Unconjugated | |
Rabbit | |
200 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot, Western Blot | |
Conc. Not Determined | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
This Histone H2A/H4 phospho Ser1 antibody was raised against a peptide including phospho-serine 1 of human histone H2A. | |
RUO | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction