Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A Rabbit anti-Human, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Invitrogen MA524662

Catalog No. PIMA524662


Add to cart

Description

Description

This antibody reacts to the Histone H2A protein, independent of post-translational modifications. No cross reactivity with other histone proteins. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using Recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
Specifications

Specifications

Histone H2A
Recombinant Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4
P04908, P0C0S8, P20671, Q96KK5, Q99878
H2A histone family, member R; histone 1, H2aa; Histone H2A type 1-A; Histone H2A/r
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3013, 8331, 8335, 85235, 8969
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
RM225
Unconjugated
HIST1H2AG
Liquid
HIST1H2AB, HIST1H2AD, HIST1H2AG, HIST1H2AH, HIST1H2AJ
Peptide corresponding to the C-terminus of human Histone H2A.
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only