Learn More
Histone H2a Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259261
Catalog No. NBP259261
Add to cart
Description
Description
Histone H2a Polyclonal antibody specifically detects Histone H2a in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Dot Blot, Immunofluorescence.
Specifications
Specifications
Histone H2a
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460
Rabbit
IgG
50 ug
Chromatin Research, Epigenetics, Histones and Modified Histones
Primary
8338
Western Blot, ChIP assay, ELISA, Dot Blot, Immunofluorescence
Western Blot 1:1000, Chromatin Immunoprecipitation 0.5-1 ug, ELISA 1:4000, Dot Blot 1:5000, Immunofluorescence 1:500
Affinity Purified
HIST2H2AC
The exact immunogen is propietary information.
Peptide affinity purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human