Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2A.Z, ac Lys7 Antibody (RM222), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326026AF594
Description
Histone H2A.Z Monoclonal antibody specifically detects Histone H2A.Z in Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex ImmunoassaySpecifications
Histone H2A.Z | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Vertebrates | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Multiplex | |
RM222 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay | |
H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z | |
An acetyl-peptide corresponding to Histone H2A.Z [ac Lys7] | |
0.1 mL | |
Epigenetics | |
3015 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction