Learn More
Histone H2A.z Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164411AP
Catalog No. 50-173-0945
Add to cart
Description
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
Specifications
Specifications
Histone H2A.z
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P0C0S5, P0C0S6, P0C0S7
H2A histone family, member Z, H2A.z, H2A/z, H2AFZ, H2AZ, Histone H2A.Z
Rabbit
IgG
150 μL
-20°C
Primary
3015, 51788, 58940
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
H2AFZ
Liquid
H2AFZ
Histone H2A.z Fusion Protein Ag9762
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat