Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AE, p Ser1 Antibody (RM216), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326017PCP
Description
Histone H2AE Monoclonal antibody specifically detects Histone H2AE in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex ImmunoassaySpecifications
Histone H2AE | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Vertebrates | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Multiplex | |
RM216 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay | |
H2A histone family, member M, H2A/m, H2AFA, H2AFMHistone H2A/m, HIST1H2AE, histone 1, H2ab, histone cluster 1, H2ab, histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a | |
An unmodified peptide corresponding to the N-terminus of Histone H2AE | |
0.1 mL | |
Epigenetics | |
3012 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction