Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | gamma H2AX |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Histone H2AX Polyclonal specifically detects Histone H2AX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
gamma H2AX | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
H2A.X, H2A/X, H2AFX | |
H2AFX | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
3014 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:AVRNDEELNKLLGGVTIAQGGVLPNI | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title