Learn More
Enzo Life SciencesSupplier Diversity Partner Histone H2AX (pSer139) monoclonal antibody (9F3) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMCC255F
Clone: 9F3. Host: Mouse. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to a portion of human Histone H2AX phosphorylated at Ser139. UniProt: P16104. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-666-470