Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Histone H2AX
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092667
View Documents
Novus Biologicals
NBP303951100UL
100 μg
Each for $320.71
Add to cart
 
NB092668
View Documents
Novus Biologicals
NBP30395120UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Histone H2AX Polyclonal antibody specifically detects Histone H2AX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

Histone H2AX
Unconjugated
Rabbit
Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific
PBS with 50% glycerol, pH7.3.
A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human Histone H2AX (NP_002096.1).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse, Rat
3014
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit