Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2AY/macroH2A.1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Histone H2AY/macroH2A.1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15300320
|
Novus Biologicals
NBP15300320UL |
20 μL |
Each for $152.22
|
|
NBP153003
|
Novus Biologicals
NBP153003 |
100 μL |
Each for $436.00
|
|
Description
Histone H2AY/macroH2A.1 Polyclonal specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Western Blot.Specifications
Histone H2AY/macroH2A.1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 | |
H2AFY | |
IgG | |
Affinity Purified | |
39 kDa |
Western Blot | |
Unconjugated | |
RUO | |
O75367 | |
9555 | |
Synthetic peptides corresponding to H2AFY(H2A histone family, member Y) The peptide sequence was selected from the N terminal of H2AFY. Peptide sequence HPKYRIGVGAPVYMAAVLEYLTAEILELAGNAARDNKKGRVTPRHILLAV. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title