Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2B, member S Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 13 publications
$206.50 - $499.50
Specifications
Antigen | Histone H2B, member S |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml, Immunoblotting reported in scientific literature (PMID 28235947) |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoblot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056347T
![]() |
Novus Biologicals
NB10056347SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056347
![]() |
Novus Biologicals
NB10056347 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Histone H2B, member S Polyclonal specifically detects Histone H2B, member S in Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammal, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
Histone H2B, member S | |
Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml, Immunoblotting reported in scientific literature (PMID 28235947) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammalia, Zebrafish | |
H2B histone family S member10H2B/s, H2B histone family, member S, Histone H2B.s | |
H2BFS | |
IgG | |
Protein G purified | |
A 15 kDa band is observed. This antibody is routinely quality controlled in human lysates. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), Immunoblot | |
Unconjugated | |
Rabbit | |
Chromatin Research | |
PBS with 0.05% Sodium Azide | |
54145 | |
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acid residues 111-125 AVSEGTKAVTKYTSS of human Histone H2B; GenBank no. CAI19747.1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title