Learn More
Histone H2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158571AP
Catalog No. 50-173-0534
Add to cart
Description
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transcription regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications
Specifications
Histone H2B
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16778, Q64524
GL105, H2B, H2B.1, H2B/q, H2BFQ, HIST2H2BE, histone cluster 2, H2be, Histone H2B, Histone H2B GL105, Histone H2B type 2 E, Histone H2B.q
Rabbit
IgG
150 μL
-20°C
Primary
295274, 319190, 8349
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HIST2H2BE
Liquid
Hist2h2be
Histone H2B Fusion Protein Ag7811
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat