Learn More
Histone H2B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259272
Catalog No. NBP259272
Add to cart
Description
Description
Histone H2B ac Lys15 Polyclonal antibody specifically detects Histone H2B ac Lys15 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Dot Blot, Immunofluorescence.
Specifications
Specifications
Histone H2B
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Rabbit
IgG
50 ug
Chromatin Research, Epigenetics, Histones and Modified Histones
Primary
440689
Western Blot, ChIP assay, ELISA, Dot Blot, Immunofluorescence
Western Blot 1:500, Chromatin Immunoprecipitation 2 ug, ELISA 1:1000, Dot Blot 1:20000, Immunofluorescence 1:500
Affinity Purified
HIST2H2BF
The exact immunogen is propietary information.
Peptide affinity purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human