Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2BK16ac Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
Supplier: Active Motif 39122
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.Specifications
Histone H2BK16ac | |
Polyclonal | |
Unconjugated | |
H2BC12 | |
2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K16; Acetyl-Histone H2B Lys16; AV127319; dJ193B12.2; dJ221C16.3; dJ221C16.8; GL105; H2B; H2b 613; H2B GL105; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B K; H2B.1; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/s; H2B/t; H2b-613; H2BC10; H2BC4; H2BC6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; H2bfs; H2BFT; H2BGL105; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRIP1; Hist1h2ba; Hist1h2bc; Hist1h2be; HIST1H2BF; Hist1h2bg; HIST1H2BI; Hist1h2bk; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; Hist2h2be; HIST2H2BF; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, | |
Rabbit | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay, Dot Blot, Immunofluorescence, Western Blot | |
Conc. Not Determined | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q99877 | |
H2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8 | |
This Histone H2B acetyl Lys16 antibody was raised against a peptide including acetyl-lysine 16 of human histone H2B. | |
RUO | |
337873, 337874, 440689, 8339, 8341, 8346, 8347, 8348, 8349, 85236 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction