Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2BK16ac Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $546.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen Histone H2BK16ac
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3892 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39122
10 μL
Each for $135.30
Add to cart
 
50-199-3891 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
39121
200 μL
Each for $546.00
Add to cart
 
Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications

Specifications

Polyclonal
Primary
Histone H2BK16ac
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit