Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3.3 Rabbit, Clone: RM190, Abnova™

Rabbit monoclonal antibody raised against synthetic peptide of human Histone H3.3.

Manufacturer:  Abnova Corporation MAB12805

Catalog No. 89-934-251


Add to cart

Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene contains introns and its mRNA is polyadenylated, unlike most histone genes. The protein encoded is a replication-independent member of the histone H3 family. [provided by RefSeq]

Specifications

Specifications

Histone H3.3
RM190
Rabbit monoclonal antibody raised against synthetic peptide of human Histone H3.3.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
H3.3A/H3F3/MGC87782/MGC87783
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to Histone H3.3, independent of post-translational modifications. No cross reactivity with Histone H3.1 or other histone proteins.
ELISA, Western Blot
Unconjugated
ELISA (0.2 ug/mL-1 ug/mL) Western Blot (0.5 ug/mL-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
H3F3A
A synthetic peptide corresponding to Histone H3.3.
Protein A purification
RUO
Primary
3020
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit