Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3.3A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298768PCP
Description
Histone H3.3A Polyclonal antibody specifically detects Histone H3.3A in Human samples. It is validated for Western BlotSpecifications
Histone H3.3A | |
Polyclonal | |
Western Blot | |
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Histone H3.3A. | |
0.1 mL | |
Chromatin Research, Epigenetics, Phospho Specific | |
3020 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction