Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500171R
Description
Histone H3 Polyclonal antibody specifically detects Histone H3 in Human, Mouse, Rat, Insect, Xenopus, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunoblotSpecifications
Histone H3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Insect, Xenopus species, Yeast | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
DyLight 550 | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
This Histone H3 antibody was raised against a synthetic peptide made to an C-terminal portion of the human Histone H3 protein (between residues 100-136) [UniProt P68431] | |
0.1 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction